Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-methyl-1,3,4-thiadiazole, 99%, Thermo Scientific™

Click to view available options
Quantity:
50g
250g
29490-19-5
C3H4N2S2
132.20
MFCD00038349
FPVUWZFFEGYCGB-UHFFFAOYSA-N
2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
1810203
5-methyl-3H-1,3,4-thiadiazole-2-thione
CC1=NNC(=S)S1
29490-19-5
C3H4N2S2
132.20
MFCD00038349
FPVUWZFFEGYCGB-UHFFFAOYSA-N
2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
1810203
5-methyl-3H-1,3,4-thiadiazole-2-thione
CC1=NNC(=S)S1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 182°C to 185°C
Beilstein 606663
Quantity 250g
Formula Weight 132.21
Percent Purity 99%
Chemical Name or Material 2-Mercapto-5-methyl-1,3,4-thiadiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 249-667-1

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only