Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Mercapto-5-nitrobenzimidazole, 97%

Click to view available options
Quantity:
5g
25g
100g
6325-91-3
C7H5N3O2S
195.196
MFCD00804727
YPXQSGWOGQPLQO-UHFFFAOYSA-N
2-mercapto-5-nitrobenzimidazole,5-nitro-2-benzimidazolethiol,5-nitro-2-mercaptobenzimidazole,5-nitro-1h-benzo d imidazole-2-thiol,2-benzimidazolethiol, 5-nitro,2-benzimidazolinethione, 5-nitro,2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro,6-nitro-1h-benzimidazole-2-thiol,5-nitro-1h-benzimidazole-2-thiol,5-nitro-3-hydrobenzimidazole-2-thione
3717598
5-nitro-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1[N+](=O)[O-])NC(=S)N2
6325-91-3
C7H5N3O2S
195.196
MFCD00804727
YPXQSGWOGQPLQO-UHFFFAOYSA-N
2-mercapto-5-nitrobenzimidazole,5-nitro-2-benzimidazolethiol,5-nitro-2-mercaptobenzimidazole,5-nitro-1h-benzo d imidazole-2-thiol,2-benzimidazolethiol, 5-nitro,2-benzimidazolinethione, 5-nitro,2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro,6-nitro-1h-benzimidazole-2-thiol,5-nitro-1h-benzimidazole-2-thiol,5-nitro-3-hydrobenzimidazole-2-thione
3717598
5-nitro-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1[N+](=O)[O-])NC(=S)N2
Melting Point ~273°C (decomposition)
Quantity 5g
Beilstein 172127
Formula Weight 195.2
Percent Purity 97%
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA