Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-nitrobenzimidazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6325-91-3
C7H5N3O2S
195.196
MFCD00804727
YPXQSGWOGQPLQO-UHFFFAOYSA-N
2-mercapto-5-nitrobenzimidazole,5-nitro-2-benzimidazolethiol,5-nitro-2-mercaptobenzimidazole,5-nitro-1h-benzo d imidazole-2-thiol,2-benzimidazolethiol, 5-nitro,2-benzimidazolinethione, 5-nitro,2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro,6-nitro-1h-benzimidazole-2-thiol,5-nitro-1h-benzimidazole-2-thiol,5-nitro-3-hydrobenzimidazole-2-thione
3717598
5-nitro-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1[N+](=O)[O-])NC(=S)N2
6325-91-3
C7H5N3O2S
195.196
MFCD00804727
YPXQSGWOGQPLQO-UHFFFAOYSA-N
2-mercapto-5-nitrobenzimidazole,5-nitro-2-benzimidazolethiol,5-nitro-2-mercaptobenzimidazole,5-nitro-1h-benzo d imidazole-2-thiol,2-benzimidazolethiol, 5-nitro,2-benzimidazolinethione, 5-nitro,2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro,6-nitro-1h-benzimidazole-2-thiol,5-nitro-1h-benzimidazole-2-thiol,5-nitro-3-hydrobenzimidazole-2-thione
3717598
5-nitro-1,3-dihydrobenzimidazole-2-thione
C1=CC2=C(C=C1[N+](=O)[O-])NC(=S)N2
Quantity 5g
Color Brown-Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 272.0°C to 278.0°C (decomposition)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 195.2
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA