Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-7-methoxy-4-methylquinoline, 96%, Thermo Scientific™

Click to view available options
Quantity:
250mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS 52507-57-0
Melting Point 233°C to 235°C
Molecular Formula C11H11NOS
MDL Number MFCD00705021
Quantity 250mg
Synonym 7-methoxy-4-methylquinoline-2-thiol, 7-methoxy-4-methyl-quinoline-2-thiol, cbmicro_019952, cbmicro_026931, 7-methoxy-4-methylhydroquinoline-2-thione, 7-methoxy-4-methylquinoline-2 1h-thione, 2-mercapto-7-methoxy-4-methylquinoline
InChI Key QJNQZUVUCGOPFS-UHFFFAOYSA-N
SMILES CC1=CC(=S)NC2=C1C=CC(=C2)OC
IUPAC Name 7-methoxy-4-methyl-1H-quinoline-2-thione
Molecular Weight (g/mol) 205.275
PubChem CID 676907
Formula Weight 205.28
Percent Purity 96%
Chemical Name or Material 2-Mercapto-7-methoxy-4-methylquinoline
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures