Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptobenzothiazole, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
1kg
Packaging:
Glass Bottle
Plastic bottle
149-30-4
C7H5NS2
167.24
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole,2-benzothiazolethiol,captax,benzothiazolethiol,2 3h-benzothiazolethione,benzothiazole-2-thiol,1,3-benzothiazole-2-thiol,benzo d thiazole-2-thiol,rokon,rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2
149-30-4
C7H5NS2
167.24
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole,2-benzothiazolethiol,captax,benzothiazolethiol,2 3h-benzothiazolethione,benzothiazole-2-thiol,1,3-benzothiazole-2-thiol,benzo d thiazole-2-thiol,rokon,rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2
Indicator
Infrared Spectrum Authentic
Quantity 25g
Packaging Plastic bottle
Flash Point 243°C
Melting Point 172.0°C to 182.0°C
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 27, II, 233
Merck Index 15, 5935
Solubility Information yellow solution
Formula Weight 167.24
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzothiazole, 98%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves.

GHS Signal Word: Warning

EINECSNumber : 205-736-8

RTECSNumber : DL6475000

TSCA : TSCA