Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethanesulfonic Acid Sodium Salt, 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
19767-45-4
C2H5NaO3S2
164.17
XOGTZOOQQBDUSI-UHFFFAOYSA-M
sodium 2-sulfanylethane-1-sulfonate
[Na+].[O-]S(=O)(=O)CCS
19767-45-4
C2H5NaO3S2
164.17
XOGTZOOQQBDUSI-UHFFFAOYSA-M
sodium 2-sulfanylethane-1-sulfonate
[Na+].[O-]S(=O)(=O)CCS

M2494, 19767-45-4, C2H5O3S2Na

Spectrum™ 2-Mercaptoethanesulfonic Acid Sodium Salt is used therapeutically to reduce the incidence of haemorrhagic cystitis and haematuria when a patient receives ifosfamide or cyclophosphamide for cancer chemotherapy. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 5 g
CAS Max % 1
Percent Purity 98%