Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethanol, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Steel Can

Chemical Identifiers

60-24-2
C2H6OS
78.129
DGVVWUTYPXICAM-UHFFFAOYSA-N
1567
CHEBI:41218

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis and other Molecular Biology Applications
CAS 60-24-2
Packaging Steel Can
Specific Gravity 1L = 1.11kg
Molecular Formula C2H6OS
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
SMILES C(CS)O
IUPAC Name 2-sulfanylethanol
Molecular Weight (g/mol) 78.129
PubChem CID 1567
ChEBI CHEBI:41218
Quantity 500g
Formula Weight 78.13
Flash Point 74.00°C
Chemical Name or Material 2-Mercaptoethanol
Grade Ultrapure Bioreagent
Show More Show Less