Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
872-35-5
C3H4N2S
100.14
MFCD00005188
OXFSTTJBVAAALW-UHFFFAOYSA-N
2-mercaptoimidazole,1h-imidazole-2-thiol,imidazole-2-thiol,1h-imidazole-2 3h-thione,4-imidazoline-2-thione,1,3-vinylenethiourea,imidazole, 2-mercapto,2h-imidazole-2-thione, 1,3-dihydro,usaf el-57,mercaptoimidazole
1201386
1,3-dihydroimidazole-2-thione
C1=CNC(=S)N1
872-35-5
C3H4N2S
100.14
MFCD00005188
OXFSTTJBVAAALW-UHFFFAOYSA-N
2-mercaptoimidazole,1h-imidazole-2-thiol,imidazole-2-thiol,1h-imidazole-2 3h-thione,4-imidazoline-2-thione,1,3-vinylenethiourea,imidazole, 2-mercapto,2h-imidazole-2-thione, 1,3-dihydro,usaf el-57,mercaptoimidazole
1201386
1,3-dihydroimidazole-2-thione
C1=CNC(=S)N1
Melting Point 228.0°C to 231.0°C
Color Tan to White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 100.14
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 2-Mercaptoimidazole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-823-4