Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptophenol, 98+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1121-24-0
C6H6OS
126.18
MFCD00040447
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
2-hydroxythiophenol,2-mercaptophenol,o-mercaptophenol,phenol, 2-mercapto,2-hydroxy thiophenol,2-hydroxybenzenethiol,o-hydroxythiophenol,o-hydroxybenzenothiol,2-hydroxybenzenothiol,mercaptophenol
70718
2-sulfanylphenol
C1=CC=C(C(=C1)O)S
1121-24-0
C6H6OS
126.18
MFCD00040447
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
2-hydroxythiophenol,2-mercaptophenol,o-mercaptophenol,phenol, 2-mercapto,2-hydroxy thiophenol,2-hydroxybenzenethiol,o-hydroxythiophenol,o-hydroxybenzenothiol,2-hydroxybenzenothiol,mercaptophenol
70718
2-sulfanylphenol
C1=CC=C(C(=C1)O)S
Infrared Spectrum Authentic
Specific Gravity 1.22
Packaging Glass bottle
Refractive Index 1.6100 to 1.6120
Flash Point 97°C
Melting Point 20.0°C
Density 1.2200g/mL
Quantity 1g
Color Green to Yellow
Boiling Point 40.0°C (300.0 mmHg)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula SHC6H4OH
Solubility Information Solubility in water: slightly soluble.
Formula Weight 126.18
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptophenol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger