Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopropionic Acid 95%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
79-42-5
C3H6O2S
106.14
MFCD00004862
PMNLUUOXGOOLSP-UHFFFAOYSA-N
2-mercaptopropionic acid,thiolactic acid,2-mercaptopropanoic acid,2-thiolactic acid,2-thiolpropionic acid,alpha-mercaptopropionic acid,alpha-mercaptopropanoic acid,propionic acid, 2-mercapto,propanoic acid, 2-mercapto,fema no. 3180
62326
CHEBI:47872
2-sulfanylpropanoic acid
CC(C(=O)O)S
79-42-5
C3H6O2S
106.14
MFCD00004862
PMNLUUOXGOOLSP-UHFFFAOYSA-N
2-mercaptopropionic acid,thiolactic acid,2-mercaptopropanoic acid,2-thiolactic acid,2-thiolpropionic acid,alpha-mercaptopropionic acid,alpha-mercaptopropanoic acid,propionic acid, 2-mercapto,propanoic acid, 2-mercapto,fema no. 3180
62326
CHEBI:47872
2-sulfanylpropanoic acid
CC(C(=O)O)S
Refractive Index 1.4804 to 1.4824
Melting Point 10°C to 14°C
Boiling Point 102°C (16.0mmHg)
Flash Point 87°C
Specific Gravity 1.19
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.1900g/mL
Quantity 100g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3CH(SH)CO2H
Beilstein 03, 289
Fieser 15,307
Merck Index 15, 9493
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol,ether and acetone,miscible with benzol,petroleumether,chloroform,,chloroethane,diethylether
Formula Weight 106.14
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 2-Mercaptopropionic acid, 95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-206-5

RTECSNumber : UF5250000

TSCA : TSCA