Alfa Aesar™ 2-Mercaptopyridine, 98%

Manufacturer: Alfa Aesar™ A1174122

Quantity 100g

 View more versions of this product

Catalog No. AAA1174122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2637-34-5
    Molecular Formula C5H5NS
    Formula Weight 111.17
    MDL Number MFCD00006285
    Beilstein 105758
    Synonym 2-Pyridinethiol
    Chemical Name or Material 2-Mercaptopyridine

    Specifications

    Formula Weight 111.17
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2637-34-5
    Health Hazard 1 Warning
    Melting Point 124° to 130°C
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    RTECS Number UT9600000
    TSCA TSCA
    Molecular Formula C5H5NS
    MDL Number MFCD00006285
    Beilstein 105758
    Synonym 2-Pyridinethiol
    Chemical Name or Material 2-Mercaptopyridine
    Assay 98%

    Description

    Applications
    Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

    Solubility
    Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

    Notes
    Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit