Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyridine-N-oxide, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1121-31-9
C5H5NOS
127.16
MFCD00006196
YBBJKCMMCRQZMA-UHFFFAOYSA-N
pyrithione,2-mercaptopyridine n-oxide,2-mercaptopyridine-n-oxide,1-hydroxy-2-pyridinethione,omadine,2-pyridinethiol, 1-oxide,1-hydroxyl-1h-pyridine-2-thione,n-hydroxypyridine-2-thione,1-hydroxy-2 1h-pyridinethione,2-pyridinethiol 1-oxide
1570
CHEBI:36578
1-hydroxypyridine-2-thione
C1=CC(=S)N(C=C1)O
1121-31-9
C5H5NOS
127.16
MFCD00006196
YBBJKCMMCRQZMA-UHFFFAOYSA-N
pyrithione,2-mercaptopyridine n-oxide,2-mercaptopyridine-n-oxide,1-hydroxy-2-pyridinethione,omadine,2-pyridinethiol, 1-oxide,1-hydroxyl-1h-pyridine-2-thione,n-hydroxypyridine-2-thione,1-hydroxy-2 1h-pyridinethione,2-pyridinethiol 1-oxide
1570
CHEBI:36578
1-hydroxypyridine-2-thione
C1=CC(=S)N(C=C1)O
Melting Point 70°C to 73°C
Color Beige to White or Gray-Green or Green-Yellow to Yellow
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (Iodometry)
Beilstein 21, V,7, 150
Fieser 12,417; 14,268; 17,292
Merck Index 14, 7994
Formula Weight 127.16
Physical Form Powder, Crystals, and/or Chunks
Percent Purity 99%
Chemical Name or Material 2-Mercaptopyridine-N-oxide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-329-4