Alfa Aesar™ 2-Mercaptopyridine N-oxide sodium salt hydrate, 98%

Manufacturer: Alfa Aesar™ A1230314

Quantity 25g

 View more versions of this product

Catalog No. AAA1230314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 304675-78-3
    Molecular Formula C5H4NNaOS·xH2O
    Formula Weight 149.15 (Anhydrous)
    MDL Number MFCD00151244
    Merck Index 14,7994
    Chemical Name or Material 2-Mercaptopyridine N-oxide sodium salt hydrate

    Specifications

    Formula Weight 149.15 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,7994
    Health Hazard 1 Danger
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    CAS 304675-78-3
    RTECS Number UT9000000
    Molecular Formula C5H4NNaOS·xH2O
    MDL Number MFCD00151244
    Chemical Name or Material 2-Mercaptopyridine N-oxide sodium salt hydrate
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit