Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptothiazole 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
25g
82358-09-6
C3H3NS2
117.184
MFCD00022449
OCVLSHAVSIYKLI-UHFFFAOYSA-N
2-mercaptothiazole,2 3h-thiazolethione,2-thiazolethiol,1,3-thiazole-2-thiol,thiazoline-2-thione,2-mercapto-1,3-thiazole,3h-thiazole-2-thione,thiazole-2-thiol,1,3-thiazole-2 3h-thione,.delta.4-thiazoline-2-thione
1201387
3H-1,3-thiazole-2-thione
C1=CSC(=S)N1
82358-09-6
C3H3NS2
117.184
MFCD00022449
OCVLSHAVSIYKLI-UHFFFAOYSA-N
2-mercaptothiazole,2 3h-thiazolethione,2-thiazolethiol,1,3-thiazole-2-thiol,thiazoline-2-thione,2-mercapto-1,3-thiazole,3h-thiazole-2-thione,thiazole-2-thiol,1,3-thiazole-2 3h-thione,.delta.4-thiazoline-2-thione
1201387
3H-1,3-thiazole-2-thione
C1=CSC(=S)N1
Quantity 25g
Color Yellow
Melting Point 80°C
Formula Weight 117.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Mercaptothiazole