Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-3-pyridinecarboxaldehyde, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
71255-09-9
C7H7NO2
137.14
PIFFMIDNNWOQLK-UHFFFAOYSA-N
2-methoxynicotinaldehyde,2-methoxy-3-pyridinecarboxaldehyde,2-methoxy-pyridine-3-carbaldehyde,2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 2-methoxy,2-methoxy-3-pyridine carboxaldehyde,3-pyridinecarboxaldehyde,2-methoxy,2-methoxy-3-pyridinecarbaldehyde,pubchem15124,2-methoxy-3-formyl-pyridine
12533391
2-methoxypyridine-3-carbaldehyde
COC1=C(C=CC=N1)C=O
71255-09-9
C7H7NO2
137.14
PIFFMIDNNWOQLK-UHFFFAOYSA-N
2-methoxynicotinaldehyde,2-methoxy-3-pyridinecarboxaldehyde,2-methoxy-pyridine-3-carbaldehyde,2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 2-methoxy,2-methoxy-3-pyridine carboxaldehyde,3-pyridinecarboxaldehyde,2-methoxy,2-methoxy-3-pyridinecarbaldehyde,pubchem15124,2-methoxy-3-formyl-pyridine
12533391
2-methoxypyridine-3-carbaldehyde
COC1=C(C=CC=N1)C=O
Melting Point 25°C
Boiling Point 200°C to 201°C
Color Colorless to Yellow
Flash Point 96°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 137.14
Physical Form Liquid or Low Melting Solid
Percent Purity 96%
Chemical Name or Material 2-Methoxy-3-pyridinecarboxaldehyde
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning