Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H31652MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31652MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

C10H5F6NO
MFCD11519321
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile
269.14
UN3439

Specifications

46° to 48°C
C10H5F6NO
MFCD11519321
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile
250mg
269.14
UN3439
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit