Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-4-pyridinecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
105596-63-2
C7H7NO3
153.14
DPNDWFVORIGXQO-UHFFFAOYSA-N
2-methoxyisonicotinic acid,2-methoxy-4-pyridinecarboxylic acid,2-methoxy-isonicotinic acid,4-pyridinecarboxylic acid, 2-methoxy,2-methoxy-isonicotinicacid,2-methoxy-4-picolinic acid,pubchem9293,ksc179m4j,4-pyridinecarboxylicacid, 2-methoxy,2-methyloxy-4-pyridinecarboxylic acid
819942
2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1)C(=O)O
105596-63-2
C7H7NO3
153.14
DPNDWFVORIGXQO-UHFFFAOYSA-N
2-methoxyisonicotinic acid,2-methoxy-4-pyridinecarboxylic acid,2-methoxy-isonicotinic acid,4-pyridinecarboxylic acid, 2-methoxy,2-methoxy-isonicotinicacid,2-methoxy-4-picolinic acid,pubchem9293,ksc179m4j,4-pyridinecarboxylicacid, 2-methoxy,2-methyloxy-4-pyridinecarboxylic acid
819942
2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1)C(=O)O
Melting Point 186°C to 188°C
Packaging Glass Bottle
Quantity 1g
Color Brown to White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 153.14
Physical Form Solid or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-pyridinecarboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning