Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-methylaniline, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
120-71-8
C8H11NO
137.18
MFCD00007815
WXWCDTXEKCVRRO-UHFFFAOYSA-N
p-cresidine,cresidine,5-methyl-o-anisidine,para-cresidine,benzenamine, 2-methoxy-5-methyl,krezidin,2-amino-4-methylanisole,3-amino-4-methoxytoluene,4-methoxy-m-toluidine,6-methoxy-m-toluidine
8445
CHEBI:82307
2-methoxy-5-methylaniline
CC1=CC(=C(C=C1)OC)N
120-71-8
C8H11NO
137.18
MFCD00007815
WXWCDTXEKCVRRO-UHFFFAOYSA-N
p-cresidine,cresidine,5-methyl-o-anisidine,para-cresidine,benzenamine, 2-methoxy-5-methyl,krezidin,2-amino-4-methylanisole,3-amino-4-methoxytoluene,4-methoxy-m-toluidine,6-methoxy-m-toluidine
8445
CHEBI:82307
2-methoxy-5-methylaniline
CC1=CC(=C(C=C1)OC)N
Color Brown to Black
Flash Point 111°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 235.0°C
Melting Point 49.0°C to 55.0°C
Quantity 25g
Density 111g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3OC6H3(CH3)NH2
Beilstein 13, II, 338
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 137.18
Physical Form Crystalline Chunks
Percent Purity 95%
Chemical Name or Material 2-Methoxy-5-methylaniline
Show More Show Less

GHS H Statement:
May cause cancer.
Harmful if swallowed.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger