Alfa Aesar™ 2-Methoxy-5-methylbenzeneboronic acid, 95%

Manufacturer: Alfa Aesar™ H5305903

Quantity 1g

 View more versions of this product

Catalog No. AAH5305903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127972-00-3
    Molecular Formula C8H11BO3
    Formula Weight 165.99
    MDL Number MFCD02179465
    Chemical Name or Material 2-Methoxy-5-methylbenzeneboronic acid

    Specifications

    Formula Weight 165.99
    Molecular Formula C8H11BO3
    MDL Number MFCD02179465
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number Y
    Melting Point 80° to 84°C
    Quantity 1g
    CAS 127972-00-3
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Methoxy-5-methylbenzeneboronic acid
    Assay 95%

    Description

    Applications
    Reactant for coupling reactions, Preparation of selective quinazolinyl-phenol inhibitors of CHK1 as potential antitumors and radioprotectants, Rh-catalyzed asymmetric arylation of aldehydes, Stereoselective hydroarylation reactions, Suzuki-Miyaura cross-coupling.

    Solubility
    Soluble in methanol.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit