Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-methylphenyl isothiocyanate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
190774-56-2
C9H9NOS
179.2
MFCD00041083
CYKFAZDMLSXWKZ-UHFFFAOYSA-N
2-methoxy-5-methylphenyl isothiocyanate,2-methoxy-5-methylphenylisothiocyanate,benzene,2-isothiocyanato-1-methoxy-4-methyl,acmc-20anrv,2-methoxy-5-methylbenzenisothiocyanate,2-isothiocyanato-1-methoxy-4-methyl-benzene,#,benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
601672
2-isothiocyanato-1-methoxy-4-methylbenzene
CC1=CC(=C(C=C1)OC)N=C=S
190774-56-2
C9H9NOS
179.2
MFCD00041083
CYKFAZDMLSXWKZ-UHFFFAOYSA-N
2-methoxy-5-methylphenyl isothiocyanate,2-methoxy-5-methylphenylisothiocyanate,benzene,2-isothiocyanato-1-methoxy-4-methyl,acmc-20anrv,2-methoxy-5-methylbenzenisothiocyanate,2-isothiocyanato-1-methoxy-4-methyl-benzene,#,benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
601672
2-isothiocyanato-1-methoxy-4-methylbenzene
CC1=CC(=C(C=C1)OC)N=C=S
Boiling Point 130°C (0.5mmHg)
Specific Gravity 1.14
Flash Point >110°C
Density 1.1400g/mL
Refractive Index 1.6290 to 1.631
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 179.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-methylphenyl isothiocyanate, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger