Alfa Aesar™ 2-Methoxy-5-nitroaniline, 98+%

Manufacturer: Alfa Aesar™ B2559114

Quantity 25g

 View more versions of this product

Catalog No. AAB2559114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99-59-2
    Molecular Formula C7H8N2O3
    Formula Weight 168.15
    MDL Number MFCD00007261
    Beilstein 879620
    Synonym 2-Amino-4-nitroanisole; 5-Nitro-o-anisidine
    Chemical Name or Material 2-Methoxy-5-nitroaniline

    Specifications

    Formula Weight 168.15
    MDL Number MFCD00007261
    Melting Point 117° to 119°C
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 99-59-2
    Molecular Formula C7H8N2O3
    Synonym 2-Amino-4-nitroanisole; 5-Nitro-o-anisidine
    Health Hazard 2 GHS H Statement
    H301-H311-H351-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Suspected of causing cancer.
    Harmful if inhaled.
    RTECS Number BZ7175000
    Beilstein 879620
    Health Hazard 1 Danger
    Chemical Name or Material 2-Methoxy-5-nitroaniline
    Assay ≥98%

    GHS H Statement
    H301-H311-H351-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Suspected of causing cancer.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit