Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxy-5-nitroaniline, 98+%

Manufacturer:  Alfa Aesar™ B2559122

Quantity 100g

 View more versions of this product

Catalog No. AAB2559122


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

99-59-2
168.15
879620
2-Methoxy-5-nitroaniline
C7H8N2O3
MFCD00007261
2-Amino-4-nitroanisole; 5-Nitro-o-anisidine

Specifications

99-59-2
168.15
879620
2-Amino-4-nitroanisole; 5-Nitro-o-anisidine
2-Methoxy-5-nitroaniline
C7H8N2O3
MFCD00007261
117° to 119°C
100g
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H351-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BZ7175000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit