Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitroaniline, 99%, Acros Organics

Click to view available options
Quantity:
5g
99-59-2
C7H8N2O3
168.152
MFCD00007261
NIPDVSLAMPAWTP-UHFFFAOYSA-N
2-amino-4-nitroanisole,5-nitro-o-anisidine,azoamine scarlet k,fast scarlet r,2-methoxy-5-nitrobenzenamine,2-amino-1-methoxy-4-nitrobenzene,azoamine scarlet,5-nitro-2-methoxyaniline,o-anisidine nitrate,o-anisidine, 5-nitro
7447
CHEBI:48977
2-methoxy-5-nitroaniline
COC1=C(C=C(C=C1)[N+](=O)[O-])N
99-59-2
C7H8N2O3
168.152
MFCD00007261
NIPDVSLAMPAWTP-UHFFFAOYSA-N
2-amino-4-nitroanisole,5-nitro-o-anisidine,azoamine scarlet k,fast scarlet r,2-methoxy-5-nitrobenzenamine,2-amino-1-methoxy-4-nitrobenzene,azoamine scarlet,5-nitro-2-methoxyaniline,o-anisidine nitrate,o-anisidine, 5-nitro
7447
CHEBI:48977
2-methoxy-5-nitroaniline
COC1=C(C=C(C=C1)[N+](=O)[O-])N
Infrared Spectrum Authentic
Melting Point 116°C to 120°C
Quantity 5g
Color Brown to Orange
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OC6H3(NO2)NH2
Beilstein 13, 389
Formula Weight 168.15
Physical Form Amorphous Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Methoxy-5-nitroaniline, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-770-5

RTECSNumber : BZ7175000

TSCA : TSCA