Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
5446-92-4
C6H6N2O3
154.12
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
5446-92-4
C6H6N2O3
154.12
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 108.0°C to 111.0°C
Color Colorless to Yellow
Density 0.45g/mL
Quantity 25g
Flash Point 130°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3O(C5H3N)NO2
Beilstein 21, II, 33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,,insoluble in hexane
Formula Weight 154.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-nitropyridine, 97%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 226-661-7

TSCA : TSCA