Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitropyridine 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
5446-92-4
C6H6N2O3
154.12
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
5446-92-4
C6H6N2O3
154.12
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
Density 0.45g/mL
Melting Point 108.0°C to 111.0°C
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
Color Colorless to Yellow
Flash Point 130°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3O(C5H3N)NO2
Beilstein 21, II, 33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,,insoluble in hexane
Formula Weight 154.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-nitropyridine, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 226-661-7

TSCA : TSCA