Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitropyridine, 97%, Acros Organics™

Click to view available options
Quantity:
5g
5446-92-4
C6H6N2O3
154.125
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
5446-92-4
C6H6N2O3
154.125
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine,5-nitro-2-methoxypyridine,6-methoxy-3-nitropyridine,pyridine, 2-methoxy-5-nitro,methoxy 2--5-nitropyridine,2-methoxy-5-nitropyridin,pubchem1207,acmc-1art3,2-methoxy-5-nitro pyridine,5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=NC=C(C=C1)[N+](=O)[O-]
Quantity 5g
Infrared Spectrum Authentic
Melting Point 108.0°C to 111.0°C
Flash Point 130°C
Linear Formula CH3O(C5H3N)NO2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,, insoluble in hexane
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-nitropyridine

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning