Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A107700E

Quantity 2500g

 View more versions of this product

Catalog No. AAA107700E


  Check Availability
Add to cart

Specifications

Description

Applications
2-Methoxybenzaldehyde has been used to examine the acaricidal activity of Periploca sepium oil and its active component against Tyrophagus. It has also been used to obtain good enantioselectivities using Cu(OAc)(2)-bis(oxazolines) via hydrogen bonding in asymmetric Henry reaction.

Solubility
Insoluble in water.

Notes
It is sensitive to air. Incompatible with oxidizing agents and bases.
Description & Specifications

Chemical Identifiers

135-02-4
136.15
606301
2-Methoxybenzaldehyde
C8H8O2
MFCD00003308
o-Anisaldehyde

Specifications

117°C (242°F)
237° to 238°C
2500g
C8H8O2
MFCD00003308
o-Anisaldehyde
98%
Characteristic
35° to 39°C
135-02-4
136.15
606301
2-Methoxybenzaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : BZ2610000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit