Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzenethiol, 97%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
50mL
Packaging:
Glass bottle
7217-59-6
C7H8OS
140.2
MFCD00004834
DSCJETUEDFKYGN-UHFFFAOYSA-N
2-methoxythiophenol,thioguaiacol,benzenethiol, 2-methoxy,o-methoxythiophenol,o-methoxybenzenethiol,2-methoxy thiophenol,2-mercaptoanisole,2-methoxybenzene-1-thiol,2-methoxy-benzenethiol,benzenethiol, o-methoxy
23642
2-methoxybenzenethiol
COC1=CC=CC=C1S
7217-59-6
C7H8OS
140.2
MFCD00004834
DSCJETUEDFKYGN-UHFFFAOYSA-N
2-methoxythiophenol,thioguaiacol,benzenethiol, 2-methoxy,o-methoxythiophenol,o-methoxybenzenethiol,2-methoxy thiophenol,2-mercaptoanisole,2-methoxybenzene-1-thiol,2-methoxy-benzenethiol,benzenethiol, o-methoxy
23642
2-methoxybenzenethiol
COC1=CC=CC=C1S
Specific Gravity 1.14
Refractive Index 1.5905 to 1.5925
Quantity 1mL
Flash Point 95°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 225.0°C to 228.0°C
Density 1.1400g/mL
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3OC6H4SH
Solubility Information Solubility in water: insoluble in water
Formula Weight 140.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzenethiol, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-605-7

RTECSNumber : DC1790000

TSCA : TSCA