Alfa Aesar™ 2-Methoxybenzophenone, 98%

Manufacturer: Alfa Aesar™ B2337603

Quantity 1g

 View more versions of this product

Catalog No. AAB2337603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2553-04-0
    Molecular Formula C14H12O2
    Formula Weight 212.25
    MDL Number MFCD00017163
    Beilstein 1873339
    Chemical Name or Material 2-Methoxybenzophenone

    Specifications

    Formula Weight 212.25
    CAS 2553-04-0
    Quantity 1g
    Molecular Formula C14H12O2
    MDL Number MFCD00017163
    Beilstein 1873339
    Boiling Point 149° to 150°C (1mmHg)
    Melting Point 36° to 38°C
    Chemical Name or Material 2-Methoxybenzophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit