Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxybenzophenone, 98%

Click to view available options
Quantity:
1g
5g
2553-04-0
C14H12O2
212.248
MFCD00017163
CSUUDNFYSFENAE-UHFFFAOYSA-N
2-methoxybenzophenone,o-methoxybenzophenone,2-methoxyphenyl phenyl methanone,benzophenone, 2-methoxy,methanone, 2-methoxyphenyl phenyl,2-methoxyphenyl phenyl ketone,methoxybenzophenone,acmc-1cotk,maybridge1_005821,2-methoxyphenyl-phenylmethanone
75702
(2-methoxyphenyl)-phenylmethanone
COC1=CC=CC=C1C(=O)C2=CC=CC=C2
2553-04-0
C14H12O2
212.248
MFCD00017163
CSUUDNFYSFENAE-UHFFFAOYSA-N
2-methoxybenzophenone,o-methoxybenzophenone,2-methoxyphenyl phenyl methanone,benzophenone, 2-methoxy,methanone, 2-methoxyphenyl phenyl,2-methoxyphenyl phenyl ketone,methoxybenzophenone,acmc-1cotk,maybridge1_005821,2-methoxyphenyl-phenylmethanone
75702
(2-methoxyphenyl)-phenylmethanone
COC1=CC=CC=C1C(=O)C2=CC=CC=C2
Boiling Point 149°C to 150°C (1mmHg)
Melting Point 36°C to 38°C
Quantity 5g
Beilstein 1873339
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 2-Methoxybenzophenone