Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
612-16-8
C8H10O2
138.17
MFCD00004611
WYLYBQSHRJMURN-UHFFFAOYSA-N
2-methoxybenzyl alcohol,2-methoxyphenyl methanol,o-methoxybenzyl alcohol,benzenemethanol, 2-methoxy,o-anisyl alcohol,2-methoxybenzylalcohol,2-methoxybenzenemethanol,unii-457nq8ngcg,457nq8ngcg,2-methoxyphenyl methan-1-ol
69154
(2-methoxyphenyl)methanol
COC1=CC=CC=C1CO
612-16-8
C8H10O2
138.17
MFCD00004611
WYLYBQSHRJMURN-UHFFFAOYSA-N
2-methoxybenzyl alcohol,2-methoxyphenyl methanol,o-methoxybenzyl alcohol,benzenemethanol, 2-methoxy,o-anisyl alcohol,2-methoxybenzylalcohol,2-methoxybenzenemethanol,unii-457nq8ngcg,457nq8ngcg,2-methoxyphenyl methan-1-ol
69154
(2-methoxyphenyl)methanol
COC1=CC=CC=C1CO
Refractive Index 1.5460 to 1.548
Density 1.0300g/mL
Flash Point >112°C
Boiling Point 248°C to 250°C
Specific Gravity 1.03
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250mL
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3OC6H4CH2OH
Beilstein 06, 893
Formula Weight 138.17
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Methoxybenzyl alcohol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.