Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF

Manufacturer:  Alfa Aesar™ H54177AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH54177AE


  Check Availability
Add to cart

Specifications

Description

Applications
2-Methoxybenzylmagnesium chloride is a Grignard reagent used in organic synthesis. It is involved in the synthesis of (-)-Chamobtusin A. It is also used in the preparation of molybdenum and tungsten Alkylidene complexes having a tert-butylimido ligand.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with water, oxidizing agents and bases. Air sensitive.
Description & Specifications

Chemical Identifiers

480438-46-8
180.92
UN2924
C8H9ClMgO
MFCD01319897
2-Methoxybenzylmagnesium chloride

Specifications

0.25M in 2-MeTHF
480438-46-8
180.92
UN2924
100mL
C8H9ClMgO
MFCD01319897
2-Methoxybenzylmagnesium chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H318-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit