Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methoxyestradiol, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Apoptotic, antiproliferative and antiangiogenic agent, in vitro and in vivo; acts via an estrogen receptor-independent mechanism. Induces p53-induced apoptosis via two pathways: activation of p38 and NF-κB; and activation of JNK and AP-1 leading to Bcl-2 phosphorylation. Also upregulates death receptor 5 and binds to tubulin, inhibiting its assembly.
Specifications
Quantity | 10 mg |
Formula Weight | 302.41 |
Percent Purity | >99% |
Chemical Name or Material | 2-Methoxyestradiol |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction