Learn More
2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals
Description
2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -85°C |
Color | Colorless |
Density | 0.965 |
pH | 4 to 7 |
Boiling Point | 124°C to 125°C |
Flash Point | 46°C (115°F) |
Odor | Characteristic |
Refractive Index | 1.4025 |
Linear Formula | CH3OCH2CH2OH |
Quantity | 4 L |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H360FD-H370-H373 |
Hazard Statement | GHS H Statement H360-H226-H302-H312-H332; May damage fertility or the unborn child. Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P307+P311-P312-P330-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER |
EINECSNumber | 203-713-7 |
RTECSNumber | KL5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.