Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, ACS, 99%3, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
109-86-4
109-86-4
Packaging Drum
Water 0.001
Titratable Acid 0.002 meq/g
Quantity 200 L
CAS Max % 1
Percent Purity 0.993
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.