Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
109-86-4
C3H8O2
76.095
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve,ethanol, 2-methoxy,ethylene glycol monomethyl ether,methyl oxitol,2-methoxy-1-ethanol,methoxyethanol,3-oxa-1-butanol,egme,monomethyl glycol,dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO
109-86-4
C3H8O2
76.095
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve,ethanol, 2-methoxy,ethylene glycol monomethyl ether,methyl oxitol,2-methoxy-1-ethanol,methoxyethanol,3-oxa-1-butanol,egme,monomethyl glycol,dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO

High quality chemicals for laboratory and specialized industrial use

Flash Point 39.00°C
Specific Gravity 1L = 0.96kg
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 76.1
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material 2-Methoxyethanol