Alfa Aesar™ 2-Methoxyethanol, Spectrophotometric Grade, 99% min

Manufacturer: Alfa Aesar™ 32444K7

Quantity 4L

 View more versions of this product

Catalog No. AA32444K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-86-4
    Linear Formula CH3OCH2CH2OH
    Formula Weight 76.1
    MDL Number MFCD00002867
    UN Number UN1188
    Beilstein 1731074
    Merck Index 14,6038
    Synonym Ethylene glycol monomethyl ether; Methyl Cellosolve
    Chemical Name or Material 2-Methoxyethanol
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 76.1
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    RTECS Number KL5775000
    UN Number UN1188
    Beilstein 1731074
    Refractive Index 1.4025
    Merck Index 14,6038
    Odor Characteristic
    Quantity 4L
    Physical Form Liquid
    MDL Number MFCD00002867
    Synonym Ethylene glycol monomethyl ether; Methyl Cellosolve
    Health Hazard 1 Danger
    Boiling Point 124° to 125°C
    Linear Formula CH3OCH2CH2OH
    CAS 109-86-4
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
    Density 0.965g/mL
    Flash Point 46° (115°F)
    Color Colorless
    Melting Point −85°C
    pH 4 to 7
    TSCA TSCA
    Chemical Name or Material 2-Methoxyethanol
    Assay ≤99%
    Grade Spectrophotometric grade

    GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit