Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxyethyl chloroformate, tech. 85%

Manufacturer:  Alfa Aesar™ H25937AK

Quantity 250mL

 View more versions of this product

Catalog No. AAH25937AK


  Check Availability
Add to cart

Specifications

Description

Applications
2-methoxyethyl chloroformate was used to produce di-2-dimethoxyethyl hydrazine carboxylate(alternative reagent for the mitsunobu reaction).

Solubility
Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

628-12-6
138.55
UN2742
2-Methoxyethyl chloroformate
CH3OCH2CH2CO2Cl
MFCD00058932
Chloroformic acid 2-methoxyethyl ester
Technical Grade

Specifications

59°C (138°F)
250mL
CH3OCH2CH2CO2Cl
MFCD00058932
Chloroformic acid 2-methoxyethyl ester
85%
157° to 158°C
628-12-6
138.55
UN2742
2-Methoxyethyl chloroformate
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H330-H314-H318-H226
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit