Alfa Aesar™ 2-Methoxyethyl chloroformate, tech. 85%

Manufacturer: Alfa Aesar™ H25937AC

Quantity 25mL

 View more versions of this product

Catalog No. AAH25937AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 628-12-6
    Molecular Formula CH3OCH2CH2CO2Cl
    Formula Weight 138.55
    MDL Number MFCD00058932
    UN Number UN2742
    Synonym Chloroformic acid 2-methoxyethyl ester
    Chemical Name or Material 2-Methoxyethyl chloroformate
    Grade Technical Grade

    Specifications

    Formula Weight 138.55
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 157° to 158°C
    Quantity 25mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.
    Flash Point 59°C (138°F)
    Molecular Formula CH3OCH2CH2CO2Cl
    MDL Number MFCD00058932
    CAS 628-12-6
    UN Number UN2742
    Synonym Chloroformic acid 2-methoxyethyl ester
    Chemical Name or Material 2-Methoxyethyl chloroformate
    Assay 85%
    Grade Technical Grade

    Description

    Applications
    2-methoxyethyl chloroformate was used to produce di-2-dimethoxyethyl hydrazine carboxylate(alternative reagent for the mitsunobu reaction).

    Solubility
    Soluble in most organic solvents.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit