Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxyfuran, 97%

Click to view available options
Quantity:
1g
5g
25g
25414-22-6
C5H6O2
98.101
MFCD00003227
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
furan, 2-methoxy,2-methoxy furan,unii-25y8i1p35t,2-furyl methyl ether,furan,2-methoxy,pubchem19998,acmc-209gjs,2-methoxyfuran,2-furyl methyl ether #
117476
2-methoxyfuran
COC1=CC=CO1
25414-22-6
C5H6O2
98.101
MFCD00003227
OXCGHDNCMSOEBZ-UHFFFAOYSA-N
furan, 2-methoxy,2-methoxy furan,unii-25y8i1p35t,2-furyl methyl ether,furan,2-methoxy,pubchem19998,acmc-209gjs,2-methoxyfuran,2-furyl methyl ether #
117476
2-methoxyfuran
COC1=CC=CO1
Refractive Index 1.447
Density 1.065
Quantity 1g
Flash Point 10°C (50°F)
Boiling Point 110°C to 111°C
UN Number UN3271
Beilstein 106270
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Methoxyfuran

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 246-953-8

TSCA : No

Recommended Storage : Keep cold