Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyhydroquinone, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
824-46-4
C7H8O3
140.138
MFCD00013971
LAQYHRQFABOIFD-UHFFFAOYSA-N
2-methoxyhydroquinone,methoxyhydroquinone,1,4-benzenediol, 2-methoxy,1,4-dihydroxy-2-methoxybenzene,unii-2hi6hnr5u1,2hi6hnr5u1,o-methoxyhydroquinone,2,5-dihydroxyanisole,2-mohydrop,acmc-209pp0
69988
2-methoxybenzene-1,4-diol
COC1=C(C=CC(=C1)O)O
824-46-4
C7H8O3
140.138
MFCD00013971
LAQYHRQFABOIFD-UHFFFAOYSA-N
2-methoxyhydroquinone,methoxyhydroquinone,1,4-benzenediol, 2-methoxy,1,4-dihydroxy-2-methoxybenzene,unii-2hi6hnr5u1,2hi6hnr5u1,o-methoxyhydroquinone,2,5-dihydroxyanisole,2-mohydrop,acmc-209pp0
69988
2-methoxybenzene-1,4-diol
COC1=C(C=CC(=C1)O)O
Infrared Spectrum Authentic
Melting Point 87°C to 90°C
Color Brown-Gray to Brown-Orange
Quantity 10g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 140.14
Physical Form Crystalline Powder and Chunks
Percent Purity ≥98.5%
Chemical Name or Material 2-Methoxyhydroquinone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.