Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2045-79-6
C9H13NO
151.21
MFCD00008186
WSWPCNMLEVZGSM-UHFFFAOYSA-N
2-methoxyphenethylamine,2-2-methoxyphenyl ethylamine,2-2-methoxyphenyl ethanamine,2-2-methoxy-phenyl-ethylamine,2-2-methoxyphenyl ethan-1-amine,benzeneethanamine, 2-methoxy,o-methoxyphenethylamine,pubchem23973,acmc-209fah,ortho-methoxyphenethylamine
74896
2-(2-methoxyphenyl)ethanamine
COC1=CC=CC=C1CCN
2045-79-6
C9H13NO
151.21
MFCD00008186
WSWPCNMLEVZGSM-UHFFFAOYSA-N
2-methoxyphenethylamine,2-2-methoxyphenyl ethylamine,2-2-methoxyphenyl ethanamine,2-2-methoxy-phenyl-ethylamine,2-2-methoxyphenyl ethan-1-amine,benzeneethanamine, 2-methoxy,o-methoxyphenethylamine,pubchem23973,acmc-209fah,ortho-methoxyphenethylamine
74896
2-(2-methoxyphenyl)ethanamine
COC1=CC=CC=C1CCN
Boiling Point 236°C to 237°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Specific Gravity 1.03
Flash Point >110°C
Color Brown to Yellow
Refractive Index 1.5400 to 1.542
Density 1.0300g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H4CH2CH2NH2
Beilstein 13,II,352
Formula Weight 151.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenethylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 218-066-6