Alfa Aesar™ 2-Methoxyphenylacetone, 98+%

Manufacturer: Alfa Aesar™ B2449503

Quantity 1g

 View more versions of this product

Catalog No. AAB2449503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5211-62-1
    Molecular Formula C10H12O2
    Formula Weight 164.2
    MDL Number MFCD00008770
    Beilstein 2502308
    Synonym 1-(2-Methoxyphenyl)-2-propanone; 2-Methoxybenzyl methyl ketone
    Chemical Name or Material 2-Methoxyphenylacetone

    Specifications

    Formula Weight 164.2
    Molecular Formula C10H12O2
    MDL Number MFCD00008770
    Boiling Point 128° to 131°C (11mmHg)
    CAS 5211-62-1
    Beilstein 2502308
    Synonym 1-(2-Methoxyphenyl)-2-propanone; 2-Methoxybenzyl methyl ketone
    Quantity 1g
    Flash Point >110°C (230°F)
    Chemical Name or Material 2-Methoxyphenylacetone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit