Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenylacetone, 98%, Acros Organics

Click to view available options
Quantity:
25g
5211-62-1
C10H12O2
164.204
MFCD00008770
GMBFNZCPZFVKAT-UHFFFAOYSA-N
2-methoxyphenylacetone,1-2-methoxyphenyl propan-2-one,1-2-methoxyphenyl acetone,o-anisylacetone,o-methoxy phenyl acetone,2-acetonylanisole,o-methoxyphenylaceton,2-propanone, 1-2-methoxyphenyl,2-methoxyphenyl acetone,pubchem23387
78887
1-(2-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=CC=C1OC
5211-62-1
C10H12O2
164.204
MFCD00008770
GMBFNZCPZFVKAT-UHFFFAOYSA-N
2-methoxyphenylacetone,1-2-methoxyphenyl propan-2-one,1-2-methoxyphenyl acetone,o-anisylacetone,o-methoxy phenyl acetone,2-acetonylanisole,o-methoxyphenylaceton,2-propanone, 1-2-methoxyphenyl,2-methoxyphenyl acetone,pubchem23387
78887
1-(2-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=CC=C1OC
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 25g
Density 1.054g/mL
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4CH2COCH3
Beilstein 08, III, 397
Formula Weight 164.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methoxyphenylacetone, 98%
Show More Show Less