Alfa Aesar™ 2-Methoxyphenylacetonitrile, 98%

Manufacturer: Alfa Aesar™ B2440018

Quantity 50g

 View more versions of this product

Catalog No. AAB2440018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7035-03-2
    Molecular Formula C9H9NO
    Formula Weight 147.18
    MDL Number MFCD00001902
    UN Number UN3439
    Beilstein 2087748
    Synonym 2-Methoxybenzyl cyanide
    Chemical Name or Material 2-Methoxyphenylacetonitrile

    Specifications

    Formula Weight 147.18
    Molecular Formula C9H9NO
    MDL Number MFCD00001902
    Boiling Point 141° to 143°C (15mmHg)
    UN Number UN3439
    Beilstein 2087748
    Melting Point 66° to 70°C
    Synonym 2-Methoxybenzyl cyanide
    Quantity 50g
    Flash Point 129°C (264°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7035-03-2
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 2-Methoxyphenylacetonitrile
    Assay 98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit