Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Methoxyphenylacetonitrile, 98%
Click to view available options
Quantity:
50g
250g
1000g
Specifications
Melting Point | 66°C to 70°C |
Flash Point | 129°C (264°F) |
Quantity | 250g |
Boiling Point | 141°C to 143°C (15mmHg) |
UN Number | UN3439 |
Beilstein | 2087748 |
Formula Weight | 147.18 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methoxyphenylacetonitrile |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 230-314-5
TSCA : No
Recommended Storage : Ambient temperatures