Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5720-06-9
C7H9BO3
151.96
MFCD00236047
ROEQGIFOWRQYHD-UHFFFAOYSA-N
2-methoxybenzeneboronic acid,2-methoxyphenyl boronic acid,o-methoxyphenylboronic acid,2-methoxyphenyl boranediol,2-boronoanisole,2-methoxypenylboronic acid,o-methoxyphenyl boronic acid,2-methoxy phenylboronic acid,2-methoxybenzene boronic acid
2733958
(2-methoxyphenyl)boronic acid
B(C1=CC=CC=C1OC)(O)O
5720-06-9
C7H9BO3
151.96
MFCD00236047
ROEQGIFOWRQYHD-UHFFFAOYSA-N
2-methoxybenzeneboronic acid,2-methoxyphenyl boronic acid,o-methoxyphenylboronic acid,2-methoxyphenyl boranediol,2-boronoanisole,2-methoxypenylboronic acid,o-methoxyphenyl boronic acid,2-methoxy phenylboronic acid,2-methoxybenzene boronic acid
2733958
(2-methoxyphenyl)boronic acid
B(C1=CC=CC=C1OC)(O)O
Color White
Melting Point 106.0°C to 111.0°C
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H4B(OH)2
Solubility Information Solubility in water:+/- 2.5% in water
Formula Weight 151.96
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Methoxyphenylboronic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA