Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate
Click to view available options
Quantity:
50 g
250 g
Description
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
Specifications
Specifications
| Density | 0.758 |
| Boiling Point | 35°C to 36°C |
| Flash Point | −29°C (−20°F) |
| Odor | Ether-like |
| Refractive Index | 1.3835 |
| Quantity | 250 g |
| UN Number | UN1993 |
| Beilstein | 1734635 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H224-H302 |
| Hazard Statement | GHS H Statement H224-H301 Extremely flammable liquid and vapour. Toxic if swallowed. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 204-125-3 |
| RTECSNumber | UD0800000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction