Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypropene, 97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
116-11-0
C4H8O
72.11
MFCD00014929
YOWQWFMSQCOSBA-UHFFFAOYSA-N
2-methoxypropene,isopropenyl methyl ether,1-propene, 2-methoxy,2-methoxy-1-propene,methyl isopropenyl ether,ether, isopropenyl methyl,propene, 2-methoxy,unii-15wbg0jt6b,2-methoxy propene,15wbg0jt6b
8300
2-methoxyprop-1-ene
CC(=C)OC
116-11-0
C4H8O
72.11
MFCD00014929
YOWQWFMSQCOSBA-UHFFFAOYSA-N
2-methoxypropene,isopropenyl methyl ether,1-propene, 2-methoxy,2-methoxy-1-propene,methyl isopropenyl ether,ether, isopropenyl methyl,propene, 2-methoxy,unii-15wbg0jt6b,2-methoxy propene,15wbg0jt6b
8300
2-methoxyprop-1-ene
CC(=C)OC
Specific Gravity 0.78
Flash Point -29°C
Boiling Point 35°C to 36°C
Refractive Index 1.3810 to 1.384
Stabilizer 0.5% max. potassium carbonate
Infrared Spectrum Authentic
Density 0.7800g/mL
Color Colorless
Quantity 250mL
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3C(OCH3)=CH2
Beilstein 01, IV, 2075
Fieser 02,230; 05,360; 11,329; 12,317
Merck Index 15, 6073
Solubility Information Solubility in water: negligible
Formula Weight 72.11
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methoxypropene, Stabilized
Show More Show Less

GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
May form explosive peroxides.

GHS P Statement
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Wash face

GHS Signal Word: Danger

EINECSNumber : 204-125-3