Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypyrazine, 95%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
3149-28-8
C5H6N2O
110.12
WKSXRWSOSLGSTN-UHFFFAOYSA-N
pyrazine, methoxy,methoxypyrazine,2-methoxy-1,4-diazine,pyrazine, 2-methoxy,2-methoxy-pyrazine,unii-ryd35t7f4t,2-methoxy pyrazine,fema no. 3302,ryd35t7f4t,methoxy-pyrazine
18467
2-methoxypyrazine
COC1=NC=CN=C1
3149-28-8
C5H6N2O
110.12
WKSXRWSOSLGSTN-UHFFFAOYSA-N
pyrazine, methoxy,methoxypyrazine,2-methoxy-1,4-diazine,pyrazine, 2-methoxy,2-methoxy-pyrazine,unii-ryd35t7f4t,2-methoxy pyrazine,fema no. 3302,ryd35t7f4t,methoxy-pyrazine
18467
2-methoxypyrazine
COC1=NC=CN=C1
Boiling Point 60°C to 61°C (29.0mmHg)
Density 1.1300g/mL
Flash Point 45°C
Specific Gravity 1.13
Quantity 5g
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Solubility Information Solubility in water: in water:(20°C) slightly soluble
Formula Weight 110.12
Percent Purity 95%
Chemical Name or Material 2-Methoxypyrazine
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 221-579-8